missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-5-propyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 1-phenyl-5-propyl-1h-pyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylicacid, 1-phenyl-5-propyl, maybridge1_003832, pubchem14158, acmc-209xta, 1-phenyl-5-propyl-1h-pyrazole-4-carboxylicacid, buttpark 36\18-32, 1h-pyrazole-4-carboxylicacid,1-phenyl-5-propyl, 1h-pyrazole-4-carboxylic acid, 1-phenyl-5-propyl |
Formula Weight | 230.27 |
Percent Purity | 97% |
Chemical Name or Material | 1-Phenyl-5-propyl-1H-pyrazole-4-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'