missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclohexanol, 97%, Thermo Scientific Chemicals

Product Code. 11409906
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1589-60-2
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00021393
InChI Key DTTDXHDYTWQDCS-UHFFFAOYSA-N
Synonym 1-phenylcyclohexanol, cyclohexanol, 1-phenyl, 1-phenyl-1-cyclohexanol, phenylcyclohexanol, cyclohexanol, phenyl, 1-phenylcyclohexanol-1, 1-phenyl-cyclohexanol, maybridge1_002485, 1-phenyl-cyclohexan-1-ol, acmc-209dj8
PubChem CID 15319
IUPAC Name 1-phenylcyclohexan-1-ol
SMILES C1CCC(CC1)(C2=CC=CC=C2)O

Specifications

Melting Point 58°C to 62°C
Boiling Point 153°C (20 mmHg)
Quantity 50 g
Beilstein 1818360
Formula Weight 176.26
Percent Purity 97%
Chemical Name or Material 1-Phenylcyclohexanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'