missing translation for 'onlineSavingsMsg'
Learn More
1-(Phenylethynyl)cyclohexanol, 99%, Thermo Scientific™ Product Code.: 11404837

1-(Phenylethynyl)cyclohexanol, 99%, Thermo Scientific™

Product Code. 11404837
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11404837

Brand: Thermo Scientific Alfa Aesar B21808.03

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20109-09-5
Molecular Formula C14H16O
Molecular Weight (g/mol) 200.281
MDL Number MFCD00060883
InChI Key LBEMYSKBSYEVAG-UHFFFAOYSA-N
Synonym 1-phenylethynyl-1-cyclohexanol, 1-phenylethynyl cyclohexanol, 1-2-phenylethynyl cyclohexan-1-ol, 1-phenylethynylcyclohexanol, acmc-1cey1, 1-phenylethynyl-cyclohexanol, 1-phenylethynyl cyclohexanol #, cyclohexanol,1-2-phenylethynyl, 1-2-phenylethynyl-1-cyclohexanol, 1-2-phenyl-ethynyl-cyclohexan-1-ol
PubChem CID 593776
IUPAC Name 1-(2-phenylethynyl)cyclohexan-1-ol
SMILES C1CCC(CC1)(C#CC2=CC=CC=C2)O

Specifications

Melting Point 60°C to 62°C
Quantity 1 g
Formula Weight 200.28
Percent Purity 99%
Chemical Name or Material 1-(Phenylethynyl)cyclohexanol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10135634 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(Phenylethynyl)cyclohexanol, 99%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.