missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylvinylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Product Code. 15409977
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143825-84-7
Molecular Formula C14H19BO2
Molecular Weight (g/mol) 230.114
MDL Number MFCD06659923
InChI Key RMGBWPMWUZSIMH-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-1-phenylvinyl-1,3,2-dioxaborolane, 1-phenylvinylboronic acid pinacol ester, 2-1-phenylethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-phenylethenyl-1,3,2-dioxaborolane, 1-phenylvinylboronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-phenylethenyl, 1-phenylvinylboronic acid pinacolester, 2-1-phenylvinyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-1-phenylvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 11031671
IUPAC Name 4,4,5,5-tetramethyl-2-(1-phenylethenyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C(=C)C2=CC=CC=C2

Specifications

Melting Point 40°C to 42°C
Quantity 250 mg
Formula Weight 230.11
Percent Purity 95%
Chemical Name or Material 1-Phenylvinylboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'