missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propanephosphonic acid cyclic anhydride, 50 wt.% solution in dimethylformamide, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 68957-94-8, 68-12-2 |
---|---|
Molecular Formula | C9H21O6P3 |
Molecular Weight (g/mol) | 318.18 |
MDL Number | MFCD00006583 |
InChI Key | PAQZWJGSJMLPMG-UHFFFAOYSA-N |
Synonym | 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide, 1-propanephosphonic acid cyclic anhydride, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphinane 2,4,6-trioxide, unii-i6egd8839n, 1,3,5,2,4,6-trioxatriphosphorinane, 2,4,6-tripropyl-, 2,4,6-trioxide, propane phosphonic acid anhydride, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane 2,4,6-trioxide, 1-propanephosphonic anhydride in thf, 2,4,6-tri-n-propyl-2,4,6-trioxo-1,3,5,2,4,6-trioxatriphosphorinane, 1-propanephosphonic anhydride in toluene, t3p |
PubChem CID | 111923 |
IUPAC Name | 2,4,6-tripropyl-1,3,5,2$l^{5},4$l^{5},6$l^{5}-trioxatriphosphinane 2,4,6-trioxide |
SMILES | CCCP1(=O)OP(=O)(OP(=O)(O1)CCC)CCC |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68957-94-8, 68-12-2 | |
318.18 | |
PAQZWJGSJMLPMG-UHFFFAOYSA-N | |
111923 | |
CCCP1(=O)OP(=O)(OP(=O)(O1)CCC)CCC |
C9H21O6P3 | |
MFCD00006583 | |
2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide, 1-propanephosphonic acid cyclic anhydride, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphinane 2,4,6-trioxide, unii-i6egd8839n, 1,3,5,2,4,6-trioxatriphosphorinane, 2,4,6-tripropyl-, 2,4,6-trioxide, propane phosphonic acid anhydride, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane 2,4,6-trioxide, 1-propanephosphonic anhydride in thf, 2,4,6-tri-n-propyl-2,4,6-trioxo-1,3,5,2,4,6-trioxatriphosphorinane, 1-propanephosphonic anhydride in toluene, t3p | |
2,4,6-tripropyl-1,3,5,2$l^{5},4$l^{5},6$l^{5}-trioxatriphosphinane 2,4,6-trioxide |
Specifications
68957-94-8, 68-12-2 | |
51.0 | |
1.0900g/mL | |
56°C | |
1.4500 to 1.4540 | |
15, 7978 | |
2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide, 1-propanephosphonic acid cyclic anhydride, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphinane 2,4,6-trioxide, unii-i6egd8839n, 1,3,5,2,4,6-trioxatriphosphorinane, 2,4,6-tripropyl-, 2,4,6-trioxide, propane phosphonic acid anhydride, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane 2,4,6-trioxide, 1-propanephosphonic anhydride in thf, 2,4,6-tri-n-propyl-2,4,6-trioxo-1,3,5,2,4,6-trioxatriphosphorinane, 1-propanephosphonic anhydride in toluene, t3p | |
PAQZWJGSJMLPMG-UHFFFAOYSA-N | |
2,4,6-tripropyl-1,3,5,2$l^{5},4$l^{5},6$l^{5}-trioxatriphosphinane 2,4,6-trioxide | |
111923 | |
≥50% solids | |
1-Propanephosphonic acid cyclic anhydride |
49.0 | |
Brown to Yellow | |
153.0°C | |
C9H21O6P3 | |
MFCD00006583 | |
1.09 | |
Solubility in water: miscible. | |
CCCP1(=O)OP(=O)(OP(=O)(O1)CCC)CCC | |
318.18 | |
318.18 | |
Solution |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May damage the unborn child.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1-Propanephosphonic acid cyclic anhydride, 50 wt.% solution in dimethylformamide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H360D-May damage the unborn child.
- H312-Harmful in contact with skin.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only