missing translation for 'onlineSavingsMsg'
Learn More

1-Propanethiol, 98%

Product Code. 11385707
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11385707

Brand: Thermo Scientific Alfa Aesar 043420.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-03-9 | C3H8S | 76.157 g/mol

1-Propanethiol, is used widely in research areas. This compound has a wide range of applications including cell biology, materials science, nanoscience, self-assembled monolayers (SAMs), nanoelectronics, and contact printing. It is also used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-03-9
Molecular Formula C3H8S
Molecular Weight (g/mol) 76.157
MDL Number MFCD00004900
InChI Key SUVIGLJNEAMWEG-UHFFFAOYSA-N
Synonym 1-propanethiol, propanethiol, n-propylmercaptan, propyl mercaptan, n-propyl mercaptan, n-propylthiol, 1-propylmercaptan, propylthiol, 1-mercaptopropane, 1-propyl mercaptan
PubChem CID 7848
ChEBI CHEBI:8473
IUPAC Name propane-1-thiol
SMILES CCCS

Specifications

Melting Point -113°C
Color Colorless
Density 0.8410 g/mL
Boiling Point 67°C to 68°C
Flash Point −20°C (4°F)
Odor Stench
Linear Formula CH3CH2CH2SH
Refractive Index 1.438
Quantity 100 mL
UN Number UN2402
Sensitivity Air Sensitive
Solubility Information It is soluble in water (1.9g/L) at 25°C. It is also soluble in alcohol,acetone,benzene,ether,hydrocarbons.
Formula Weight 76.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Propanethiol
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Propanethiol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Propanethiol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.