missing translation for 'onlineSavingsMsg'
Learn More
1-Propanol, 99.5%, for analysis, Thermo Scientific Chemicals Product Code.: 10521681

1-Propanol, 99.5%, for analysis, Thermo Scientific Chemicals

Product Code. 10521681
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10521681

Brand: Thermo Scientific Acros 232070010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127.0°C
CAS Min % 99.4
Color Beige-Yellow to White
Acidity 0.0004 meq/g max.
CAS Max % 100.0
Alkalinity 0.0006 meq/g max.
Density 0.8000g/mL
Boiling Point 97.0°C
Quantity 1 L
Flash Point 15°C
Packaging Glass Bottle
Physical Form Crystalline Mass, Powder, Crystals and/or Chunks
Refractive Index 1.3840 to 1.3860
Assay Percent Range 99.5%
Residue after Evaporation 0.001 % max.
Specific Gravity 0.8
Linear Formula CH3CH2CH2OH
Viscosity 2.2 mPa.s (20°C)
Beilstein 01, IV, 1413
Water 0.1% Max.
Merck Index 15, 7955
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents
Formula Weight 60.1
Percent Purity 99.5%
Grade Analytical
Chemical Name or Material 1-Propanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2405230 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number232070010,232070025,232070100
Lot Number1904925 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number232070010,232070025,232070100
Lot Number1989039 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number232070010,232070025,232070100
Lot Number2220622 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number232070010,232070025,232070100
Lot Number2041865 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number232070010,232070025,232070100
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Propanol, 99.5%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.