missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propanol, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
500 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Melting Point | -127°C |
Linear Formula | CH3CH2CH2OH |
UN Number | UN1274 |
Beilstein | 1098242 |
Formula Weight | 60.1g/mol |
Percent Purity | ≥99.5% |
Density | 0.804g/mL at 25°C |
Boiling Point | 97°C |
Quantity | 1 L |
Explosion Limit | 0.137 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'