missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, purum, Honeywell

Product Code. 15652510
Click to view available options
Quantity:
1 L
2.5 L
20 kg
160 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Density 0.804g/mL at 25°C
Quantity 160 kg
Explosion Limit 0.137
Flash Point 22°C (71.6°F)
Ignition Point 700°F
Linear Formula CH3CH2CH2OH
Vapor Pressure 10mmHg (147°C), 14.9mmHg (20°C)
UN Number 1274
Beilstein 1098242
Formula Weight 60.1g/mol
Percent Purity ≥99% (GC)
Grade purum
Chemical Name or Material 1-Propanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'