missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propylamine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
2500 mL
Description
1-Propylamine is used in chemical analysis and to make other chemicals, used as a solvent in organic synthesis. And it is used as finishing agent of drugs, paints, rubber, fiber, textile and resin. Additionally, it can be used as petroleum additive and preservative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Propylamine is used in chemical analysis and to make other chemicals, used as a solvent in organic synthesis. And it is used as finishing agent of drugs, paints, rubber, fiber, textile and resin. Additionally, it can be used as petroleum additive and preservative.
Solubility
Miscible with water, alcohol, ether
Notes
Miscible with water, alcohol, ether
1-Propylamine is used in chemical analysis and to make other chemicals, used as a solvent in organic synthesis. And it is used as finishing agent of drugs, paints, rubber, fiber, textile and resin. Additionally, it can be used as petroleum additive and preservative.
Solubility
Miscible with water, alcohol, ether
Notes
Miscible with water, alcohol, ether
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -83°C |
Density | 0.719 |
pH | 12.6 |
Boiling Point | 48°C to 49°C |
Flash Point | −12°C (10°F) |
Odor | Amine-like |
Refractive Index | 1.389 |
Quantity | 2500 mL |
UN Number | UN1277 |
Beilstein | 1098243 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'