missing translation for 'onlineSavingsMsg'
Learn More

1-Propylboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 11374346
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17745-45-8
Molecular Formula C3H9BO2
Molecular Weight (g/mol) 87.91
MDL Number MFCD01074564
InChI Key JAQOMSTTXPGKTN-UHFFFAOYSA-N
Synonym n-propylboronic acid, 1-propaneboronic acid, propaneboronic acid, boronic acid, propyl, 1-propylboronic acid, 1-propaneboronic, n-propylboronc acd, propyl boronic acid, pubchem6019
PubChem CID 351065
IUPAC Name propylboronic acid
SMILES CCCB(O)O

Specifications

Melting Point 105°C to 107°C
Quantity 5 g
Beilstein 1731884
Sensitivity Air sensitive; Hygroscopic
Formula Weight 87.91
Percent Purity 98%
Chemical Name or Material 1-Propylboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'