missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Pyrrolidinecarbonyl chloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 75.0°C (1.0 mmHg) |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Assay Percent Range | 96% min. (GC) |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Refractive Index | 1.4918 to 1.4938 |
Formula Weight | 133.58 |
Percent Purity | 97% |
Show More |
Safety and Handling
Product Identifier
- 1-Pyrrolidinecarbonyl chloride
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'