Learn More
Uvex™ 1 Sport White NC Shoe O2 FO SRC, Width 11
With a lightweight design, high-tech materials, and ergonomically designed outsole, the Uvex 1 Sport White NC Shoe is the ideal companion for the working day and beyond. The hydrophobic upper ensures optimized climate control, and reliably protects the wearer against liquids and other contaminants.
£98.45
Specifications
Color | White |
---|---|
Certifications/Compliance (Europe) | EN ISO 20347:2012 O2 FO SRC EN ISO 20347:2022 O2 FO SR |
Gender | Unisex |
Fit | Standard Width |
Latex Free | Yes |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17823280
|
Uvex™
6572835 |
35 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17833280
|
Uvex™
6572836 |
36 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17843280
|
Uvex™
6572837 |
37 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17853280
|
Uvex™
6572838 |
38 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17863280
|
Uvex™
6572839 |
39 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17873280
|
Uvex™
6572840 |
40 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17883280
|
Uvex™
6572841 |
41 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17893280
|
Uvex™
6572842 |
42 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17803290
|
Uvex™
6572843 |
43 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17813290
|
Uvex™
6572844 |
44 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17823290
|
Uvex™
6572845 |
45 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17833290
|
Uvex™
6572846 |
46 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17843290
|
Uvex™
6572847 |
47 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17853290
|
Uvex™
6572848 |
48 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17863290
|
Uvex™
6572849 |
49 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17873290
|
Uvex™
6572850 |
50 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17883290
|
Uvex™
6572851 |
51 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
17893290
|
Uvex™
6572852 |
52 |
£98.45
1 pair |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
Description
- Modern, ultra-lightweight and flexible O2 occupational shoe without protective toe cap
- Hydrophobic upper materials provide reliable protection against liquids and contaminants and are easy to clean
- Washable at up to 30°C
- Made from synthetic materials, so suitable for people allergic to chrome
- Elastic laces with quick release for individual adjustment
- All sole materials are free of silicone, plasticizers and other substances that interfere with wetting agents
- New, innovative uvex i-PUREnrj sole technology provides outstanding shock-absorption properties at the forefoot and heel and excellent rebound energy over the entire midsole
- Meets the ESD specification with volume resistance less than 35 Megohms
- Ergonomically designed outsole made from dual density polyurethane with excellent slip resistance
- Outstanding wearer comfort thanks to a newly developed last and climate-optimized, breathable materials
- Virtually seam-free construction with high-tech materials to eliminate pressure points
- Removable anti-static comfortable insole with a moisture transport system and additional shock absorption in the forefoot and heel
- Soft padding on collar and tongue
- Made partly from recycled materials in order to conserve resources
- Free from any of the harmful substances listed on our banned hazardous substances list
Specifications
White | |
Unisex | |
Yes |
EN ISO 20347:2012 O2 FO SRC EN ISO 20347:2022 O2 FO SR | |
Standard Width |