missing translation for 'onlineSavingsMsg'
Learn More

uvex 1 Sport White NC Slipper O2 FO SRC, Width 11

With a lightweight design, high-tech materials, and ergonomically designed outsole, the Uvex 1 Sport White NC Shoe is the ideal companion for the working day and beyond. The hydrophobic upper ensures optimized climate control, and reliably protects the wearer against liquids and other contaminants.

Brand:  uvex 6571847

77.93 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 17803240

  • £99.40 / 1 pair
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Modern, ultra-lightweight and flexible O2 occupational shoe without protective toe cap
  • Hydrophobic upper materials provide reliable protection against liquids and contaminants and are easy to clean
  • Washable at up to 30°C
  • Made from synthetic materials, so suitable for people allergic to chrome
  • All sole materials are free of silicone, plasticizers and other substances that interfere with wetting agents
  • New, innovative uvex i-PUREnrj sole technology provides outstanding shock-absorption properties at the forefoot and heel and excellent rebound energy over the entire midsole
  • Meets the ESD specification with volume resistance less than 35 Megohms
  • Ergonomically designed outsole made from dual density polyurethane with excellent slip resistance
  • Outstanding wearer comfort thanks to a newly developed last and climate-optimized, breathable materials
  • Virtually seam-free construction with high-tech materials to eliminate pressure points
  • Removable anti-static comfortable insole with a moisture transport system and additional shock absorption in the forefoot and heel
  • Soft padding on collar and tongue
  • Made partly from recycled materials in order to conserve resources
  • Free from any of the harmful substances listed on our banned hazardous substances list
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
47
EN ISO 20347:2012 O2 FO SRC EN ISO 20347:2022 O2 FO SR
Standard Width
Trainers
ESD Safety Shoe
Microfiber
Plain Toe
Unisex
Yes
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers