missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 1-Step™ Chloronaphthol Substrate Solution

1-Step™ Chloronaphthol Substrate Solution

Brand:  Thermo Scientific™ 34012


Product Code. 11809260

  • £119.00 / 250mL
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific Pierce 1-Step Chloronaphthol Solution is a solution of 4-chloro-1-naphthol for the chromogenic detection of horseradish peroxidase (HRP) activity in Western blot and tissue staining methods.

Chloronaphthol (4-CN, MW = 178.6) reacts with HRP to form a distinct blue to blue-purple product to localize and visualize the activity of peroxidase-conjugated antibodies on blots or fixed tissue samples. The HRP substrate is not as sensitive or stable as other chromogenic substrates, such as TMB and DAB. However, the blue-purple precipitate photographs well, and its alcohol-solubility is a property that can be utilized in certain double-staining applications.

Features of Chloronaphthol Substrates:

  • HRP substrate—for detection of horseradish peroxidase activity on solid media
  • Chromogenic—no special equipment needed for visualization
  • Blue-purple—easy to photograph in color or black-and-white
  • Package options—choose powder or ready-to-use solution

    Highlights of Pierce CN Solution
  • Ready-to-use—no ethanol or methanol required to dissolve; no dilution necessary for use
  • Complete—does not require buffer or addition of hydrogen peroxide
  • Pre-filtered—does not need to be filtered before use like powders or tablets after they are dissolved
  • Stable—store refrigerated at 4°C for at least one year

    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Substrate
    Liquid
    250 mL
    Horseradish Peroxidase
    Colorimetric
    1-Step™
    Horseradish Peroxidase
    Store at 4°C.
    Product Suggestions

    Product Suggestions

    Videos
    Safety and Handling

    Safety and Handling

    Product Identifier
    • 1-Step Chloronaphthol Substrate Solution

    Signal Word
    • Warning

    Hazard Category
    • Acute toxicity Category 4
    • Specific target organ toxicity Category 2

    Hazard Statement
    • H302-Harmful if swallowed.
    • H371-May cause damage to organs.

    Precautionary Statement
    • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
    • P264-Wash hands thoroughly after handling.
    • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
    • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
    • P405-Store locked up.
    • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

    Supplemental information
    • MIXTURE LIST-Contains : methanol

    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.