missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-2-indolinecarboxylic acid, 90%, Thermo Scientific™

Product Code. 10628160
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 133851-52-2
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.293
MDL Number MFCD02682411
InChI Key QONNUMLEACJFME-UHFFFAOYSA-N
Synonym 1-tert-butoxycarbonyl-2-indolinecarboxylic acid, n-boc-indoline-2-carboxylic acid, 1-boc-indoline-2-carboxylic acid, 1-tert-butoxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1-tert-butoxycarbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1-tert-butoxycarbonyl-2,3-dihydroindole-2-carboxylic acid, 1-n-boc-indoline-2-carboxylic acid, 1-2-methylpropan-2-yl oxycarbonyl-2,3-dihydroindole-2-carboxylic acid, indoline-2-carboxylic acid, n-boc protected, 1-tert-butoxycarbonyl indoline-2-carboxylicacid
PubChem CID 2794663
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]-2,3-dihydroindole-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1C(CC2=CC=CC=C21)C(=O)O

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Quantity 5 g
Formula Weight 263.29
Percent Purity 90%
Chemical Name or Material 1-(Tert-butoxycarbonyl)-2-indolinecarboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'