Learn More
1-(tert-Butoxycarbonyl)-2-indolinecarboxylic acid, 90%, Thermo Scientific™
£29.95 - £253.00
Chemical Identifiers
CAS | 133851-52-2 |
---|---|
Molecular Formula | C14H17NO4 |
Molecular Weight (g/mol) | 263.293 |
MDL Number | MFCD02682411 |
InChI Key | QONNUMLEACJFME-UHFFFAOYSA-N |
Synonym | 1-tert-butoxycarbonyl-2-indolinecarboxylic acid, n-boc-indoline-2-carboxylic acid, 1-boc-indoline-2-carboxylic acid, 1-tert-butoxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1-tert-butoxycarbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1-tert-butoxycarbonyl-2,3-dihydroindole-2-carboxylic acid, 1-n-boc-indoline-2-carboxylic acid, 1-2-methylpropan-2-yl oxycarbonyl-2,3-dihydroindole-2-carboxylic acid, indoline-2-carboxylic acid, n-boc protected, 1-tert-butoxycarbonyl indoline-2-carboxylicacid |
PubChem CID | 2794663 |
IUPAC Name | 1-[(2-methylpropan-2-yl)oxycarbonyl]-2,3-dihydroindole-2-carboxylic acid |
SMILES | CC(C)(C)OC(=O)N1C(CC2=CC=CC=C21)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10249052
|
Thermo Scientific Maybridge
MO01075CB |
250 mg |
£29.95
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10238722
|
Thermo Scientific Maybridge
MO01075DA |
1 g |
£79.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10628160
|
Thermo Scientific Maybridge
MO01075DE |
5 g |
£253.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
133851-52-2 | |
263.293 | |
QONNUMLEACJFME-UHFFFAOYSA-N | |
2794663 | |
CC(C)(C)OC(=O)N1C(CC2=CC=CC=C21)C(=O)O |
C14H17NO4 | |
MFCD02682411 | |
1-tert-butoxycarbonyl-2-indolinecarboxylic acid, n-boc-indoline-2-carboxylic acid, 1-boc-indoline-2-carboxylic acid, 1-tert-butoxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1-tert-butoxycarbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1-tert-butoxycarbonyl-2,3-dihydroindole-2-carboxylic acid, 1-n-boc-indoline-2-carboxylic acid, 1-2-methylpropan-2-yl oxycarbonyl-2,3-dihydroindole-2-carboxylic acid, indoline-2-carboxylic acid, n-boc protected, 1-tert-butoxycarbonyl indoline-2-carboxylicacid | |
1-[(2-methylpropan-2-yl)oxycarbonyl]-2,3-dihydroindole-2-carboxylic acid |
Specifications
133851-52-2 | |
90% | |
C14H17NO4 | |
1-tert-butoxycarbonyl-2-indolinecarboxylic acid, n-boc-indoline-2-carboxylic acid, 1-boc-indoline-2-carboxylic acid, 1-tert-butoxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1-tert-butoxycarbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1-tert-butoxycarbonyl-2,3-dihydroindole-2-carboxylic acid, 1-n-boc-indoline-2-carboxylic acid, 1-2-methylpropan-2-yl oxycarbonyl-2,3-dihydroindole-2-carboxylic acid, indoline-2-carboxylic acid, n-boc protected, 1-tert-butoxycarbonyl indoline-2-carboxylicacid | |
CC(C)(C)OC(=O)N1C(CC2=CC=CC=C21)C(=O)O | |
263.293 | |
263.29 | |
1-(Tert-butoxycarbonyl)-2-indolinecarboxylic acid |
90.0 | |
Amber glass bottle | |
MFCD02682411 | |
QONNUMLEACJFME-UHFFFAOYSA-N | |
1-[(2-methylpropan-2-yl)oxycarbonyl]-2,3-dihydroindole-2-carboxylic acid | |
2794663 | |
90% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.