missing translation for 'onlineSavingsMsg'
Learn More

1-[(tert-Butoxycarbonyl)amino]cyclopropanecarboxylic acid, ≥97%, Thermo Scientific™

Catalog Number 10492913
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
88950-64-5
C9H15NO4
201.22
MFCD00083257
DSKCOVBHIFAJRI-UHFFFAOYSA-N
1-boc-amino cyclopropanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropanecarboxylic acid, n-boc-1-aminocyclopropanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopropanecarboxylic acid, boc-acpc-oh, 1-tert-butoxy carbonyl amino cyclopropane-1-carboxylic acid, 1-boc amino cyclopanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 1-1,1-dimethylethoxy carbonyl amino
688396
1-{[(tert-butoxy)carbonyl]amino}cyclopropane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CC1)C(O)=O
This item is not returnable. View return policy
88950-64-5
C9H15NO4
201.22
MFCD00083257
DSKCOVBHIFAJRI-UHFFFAOYSA-N
1-boc-amino cyclopropanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropanecarboxylic acid, n-boc-1-aminocyclopropanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopropanecarboxylic acid, boc-acpc-oh, 1-tert-butoxy carbonyl amino cyclopropane-1-carboxylic acid, 1-boc amino cyclopanecarboxylic acid, 1-tert-butoxycarbonyl amino cyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 1-1,1-dimethylethoxy carbonyl amino
688396
1-{[(tert-butoxy)carbonyl]amino}cyclopropane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CC1)C(O)=O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Formula Weight 201.22
Percent Purity 97%
Quantity 250 mg
Chemical Name or Material 1-[(Tert-butoxycarbonyl)amino]cyclopropanecarboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'