missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-tert-Butyl 4-(9-H-fluoren-9-ylmethyl) hydrogen (2R)-piperazine-1,2,4-tricarboxyl, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 452.5 |
Percent Purity | 97% |
Chemical Name or Material | 1-tert-Butyl 4-(9-H-fluoren-9-ylmethyl) hydrogen (2R)-piperazine-1, 2, 4-tricarboxyl |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'