missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl 4-(pentafluorophenyl) piperidine-1,4-dicarboxylate, 97%, Thermo Scientific™

Product Code. 10507664
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 294885-28-2
Molecular Formula C17H18F5NO4
Molecular Weight (g/mol) 395.33
MDL Number MFCD09879992
InChI Key PPKHOXJANSFRFJ-UHFFFAOYSA-N
Synonym 1-tert-butyl 4-pentafluorophenyl piperidine-1,4-dicarboxylate, 1-tert-butyl 4-2,3,4,5,6-pentafluorophenyl piperidine-1,4-dicarboxylate, o1-tert-butyl o4-2,3,4,5,6-pentakis fluoranyl phenyl piperidine-1,4-dicarboxylate, 1,4-piperidinedicarboxylicacid, 1-1,1-dimethylethyl 4-2,3,4,5,6-pentafluorophenyl ester, piperidine-1,4-dicarboxylic acid o1-tert-butyl ester o4-2,3,4,5,6-pentafluorophenyl ester
PubChem CID 11749780
IUPAC Name 1-O-tert-butyl 4-O-(2,3,4,5,6-pentafluorophenyl) piperidine-1,4-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 395.32
Percent Purity 95%
Chemical Name or Material 1-tert-Butyl 4-(pentafluorophenyl) piperidine-1, 4-dicarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.