missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(tert-butyl)-5-methyl-1H-pyrazole-3-carbohydrazide, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 97% |
Quantity | 250 mg |
Formula Weight | 196.25 |
Percent Purity | 97% |
Chemical Name or Material | 1-(tert-Butyl)-5-methyl-1H-pyrazole-3-carbohydrazide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'