missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl 6-methyl 3-iodo-1H-indole-1,6-dicarboxylate, ≥97%, Thermo Scientific™

£58.70 - £159.00

Chemical Identifiers

CAS 850374-94-6
Molecular Formula C15H16INO4
Molecular Weight (g/mol) 401.2
MDL Number MFCD06797499
InChI Key KFPCBJZNHXTGHX-UHFFFAOYSA-N
Synonym 1-tert-butyl 6-methyl 3-iodo-1h-indole-1,6-dicarboxylate, 1-tert-butyl 6-methyl-3-iodo-1h-indole-1,6-dicarboxylate, 1-tert-butyl 6-methyl 3-iodoindole-1,6-dicarboxylate, 1-tert-butyl-6-methyl-3-iodo-1h-indole-1,6-dicarboxylate, methyl 3-iodo-1h-indole-6-carboxylate,n-boc protected, 1-tert-butyl-6-methyl 3-iodo-1h-indole-1,6-dicarboxylate
PubChem CID 7127827
IUPAC Name 1-O-tert-butyl 6-O-methyl 3-iodoindole-1,6-dicarboxylate
SMILES CC(C)(C)OC(=O)N1C=C(C2=C1C=C(C=C2)C(=O)OC)I
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10340213
View Documents
Thermo Scientific Maybridge
MO07419CB
250 mg Amber Glass Bottle
£58.70
250mg
Please to purchase this item. Need a web account? Register with us today!
10442722
View Documents
Thermo Scientific Maybridge
MO07419DA
1 g Amber Glass Bottle
£159.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

850374-94-6
401.2
KFPCBJZNHXTGHX-UHFFFAOYSA-N
7127827
CC(C)(C)OC(=O)N1C=C(C2=C1C=C(C=C2)C(=O)OC)I
C15H16INO4
MFCD06797499
1-tert-butyl 6-methyl 3-iodo-1h-indole-1,6-dicarboxylate, 1-tert-butyl 6-methyl-3-iodo-1h-indole-1,6-dicarboxylate, 1-tert-butyl 6-methyl 3-iodoindole-1,6-dicarboxylate, 1-tert-butyl-6-methyl-3-iodo-1h-indole-1,6-dicarboxylate, methyl 3-iodo-1h-indole-6-carboxylate,n-boc protected, 1-tert-butyl-6-methyl 3-iodo-1h-indole-1,6-dicarboxylate
1-O-tert-butyl 6-O-methyl 3-iodoindole-1,6-dicarboxylate

Specifications

850374-94-6
97%
MFCD06797499
KFPCBJZNHXTGHX-UHFFFAOYSA-N
1-O-tert-butyl 6-O-methyl 3-iodoindole-1,6-dicarboxylate
7127827
97%
97.0
C15H16INO4
1-tert-butyl 6-methyl 3-iodo-1h-indole-1,6-dicarboxylate, 1-tert-butyl 6-methyl-3-iodo-1h-indole-1,6-dicarboxylate, 1-tert-butyl 6-methyl 3-iodoindole-1,6-dicarboxylate, 1-tert-butyl-6-methyl-3-iodo-1h-indole-1,6-dicarboxylate, methyl 3-iodo-1h-indole-6-carboxylate,n-boc protected, 1-tert-butyl-6-methyl 3-iodo-1h-indole-1,6-dicarboxylate
CC(C)(C)OC(=O)N1C=C(C2=C1C=C(C=C2)C(=O)OC)I
401.2
401.19
1-(tert-Butyl)-6-methyl-3-iodo-1H-indole-1, 6-dicarboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.