missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butyldimethylsilyl)indole-5-boronic acid, 97%, Thermo Scientific™

Product Code. 15489955
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 913835-68-4
Molecular Formula C14H22BNO2Si
Molecular Weight (g/mol) 275.23
MDL Number MFCD03095156
InChI Key PPFZQIOBIOETAS-UHFFFAOYSA-N
Synonym 1-tert-butyldimethylsilyl-1h-indol-5-yl boronic acid, 1-tert-butyldimethylsilyl-1h-indol-5-ylboronic acid, 1-tert-butyldimethylsilyl-1h-indol-5-yl-5-boronic acid, 1-tert-butyldimethylsilyl indol-5-ylboronic acid, 1-tbdms-indole-5-boronic acid, 5-borono-1-tert-butyl dimethylsilyl-1h-indole, 5-borono-1h-indol-1-yl tert-butyl dimethylsilane, 1-tert-butyl dimethyl silyl indol-5-yl boronic acid, 1-tert-butyl dimethylsilyl-1h-indole-5-boronic acid, 1-tert-butyl dimethyl silyl-1h-indol-5-yl boronic acid
PubChem CID 4372773
SMILES CC(C)(C)[Si](C)(C)N1C=CC2=CC(=CC=C12)B(O)O

Specifications

Melting Point 248°C to 250°C
Quantity 250 mg
IUPAC Name [1-[tert-butyl(dimethyl)silyl]indol-5-yl]boronic acid
Formula Weight 275.23
Percent Purity 97%
Chemical Name or Material 1-(tert-Butyldimethylsilyl)indole-5-boronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'