missing translation for 'onlineSavingsMsg'
Learn More
(1-Tetradecyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals Product Code.: 11393176

(1-Tetradecyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals

Product Code. 11393176
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11393176

Brand: Thermo Scientific Alfa Aesar L12484.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4574-04-3
MDL Number MFCD00059968

Specifications

Molecular Formula C6H10O2
Quantity 25 g
Beilstein 3916572
Sensitivity Hygroscopic
Synonym cyclopropanecarboxylic acid ethyl ester, ethyl cyclopropylcarboxylate, ethyl cyclopropane carboxylate, cyclopropanecarboxylic acid, ethyl ester, pubchem11840, ethyl cylopropylcarboxylate, ethoxycarbonyl cyclopropane, ethylcyclopropane carboxylate, ksc497s5p, lddosdvzpsglfz-uhfffaoysa
InChI Key LDDOSDVZPSGLFZ-UHFFFAOYSA-N
SMILES CCOC(=O)C1CC1
IUPAC Name ethyl cyclopropanecarboxylate
Molecular Weight (g/mol) 114.14
PubChem CID 78351
Formula Weight 291.95
Percent Purity 98%
Chemical Name or Material (1-Tetradecyl)trimethylammonium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW19K025 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog NumberL12484.14,L12484.06
Lot NumberP19K006 Certificate TypeCertificate of Analysis Date12/06/2024 Catalog NumberL12484.14,L12484.06
Lot NumberW25J036 Certificate TypeCertificate of Analysis Date05/10/2023 Catalog NumberL12484.14,L12484.06
Lot NumberU18J027 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog NumberL12484.14,L12484.06
Lot NumberM16J010 Certificate TypeCertificate of Analysis Date26/01/2023 Catalog NumberL12484.14,L12484.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(1-Tetradecyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.