missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecylamine, 98%, Thermo Scientific Chemicals

£23.60 - £127.00

Chemical Identifiers

CAS 2016-42-4
Molecular Formula C14H31N
Molecular Weight (g/mol) 213.41
InChI Key PLZVEHJLHYMBBY-UHFFFAOYSA-N
Synonym tetradecylamine, 1-tetradecylamine, myristylamine, 1-tetradecanamine, n-tetradecylamine, 1-aminotetradecane, tetradecanamine, armeen 14, monotetradecylamine, alamine 5d
PubChem CID 16217
IUPAC Name tetradecan-1-amine
SMILES CCCCCCCCCCCCCCN
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10772695
View Documents
Thermo Scientific Acros
138100050
5 g Glass bottle
£23.60
5g
Please to purchase this item. Need a web account? Register with us today!
10215000
View Documents
Thermo Scientific Acros
138100100
10 g Glass bottle
£39.40
10g
Please to purchase this item. Need a web account? Register with us today!
10045723
View Documents
Thermo Scientific Acros
138100500
50 g Glass bottle
£127.00
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2016-42-4
213.41
tetradecylamine, 1-tetradecylamine, myristylamine, 1-tetradecanamine, n-tetradecylamine, 1-aminotetradecane, tetradecanamine, armeen 14, monotetradecylamine, alamine 5d
tetradecan-1-amine
C14H31N
PLZVEHJLHYMBBY-UHFFFAOYSA-N
16217
CCCCCCCCCCCCCCN

Specifications

2016-42-4
Pink to White
291°C
Authentic
C14H31N
1.5045 to 1.5065
0.993
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
CCCCCCCCCCCCCCN
213.41
213.41
Crystalline Solid or Crystals
37°C to 42°C
0.9930g/mL
>110°C
98%
CH3(CH2)13NH2
04, 201
tetradecylamine, 1-tetradecylamine, myristylamine, 1-tetradecanamine, n-tetradecylamine, 1-aminotetradecane, tetradecanamine, armeen 14, monotetradecylamine, alamine 5d
PLZVEHJLHYMBBY-UHFFFAOYSA-N
tetradecan-1-amine
16217
98%
1-Tetradecylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 217-950-9

Product Identifier
  • 1-Tetradecylamine

Signal Word
  • Danger

Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only