Learn More
1-Tetradecylphosphonic acid, 98%, Thermo Scientific Chemicals
£68.70 - £985.00
Chemical Identifiers
CAS | 4671-75-4 |
---|---|
Molecular Formula | C14H31O3P |
Molecular Weight (g/mol) | 278.373 |
MDL Number | MFCD00015834 |
InChI Key | BVQJQTMSTANITJ-UHFFFAOYSA-N |
Synonym | phosphonic acid, tetradecyl, 1-tetradecanephosphonic acid, tetradecanephosphonic acid, tetradecyl phosphonic acid, n-tetradecylphosphonic acid, tetradecyl-phosphonic acid, 1-tetradecylphosphonic acid, phosphonic acid, p-tetradecyl, p-tetradecyl-phosphonic acid, tetradecylphosphonate |
PubChem CID | 78401 |
IUPAC Name | tetradecylphosphonic acid |
SMILES | CCCCCCCCCCCCCCP(=O)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11408127
|
Thermo Scientific Alfa Aesar
A17511.03 |
1 g |
£68.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11418127
|
Thermo Scientific Alfa Aesar
A17511.06 |
5 g |
£229.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428127
|
Thermo Scientific Alfa Aesar
A17511.14 |
25 g |
£985.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Tetradecylphosphonic acid is used to functionalize the surface of BaTiO3 nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Tetradecylphosphonic acid is used to functionalize the surface of BaTiO3 nanoparticles.
Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide. Partially soluble in water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.
Chemical Identifiers
4671-75-4 | |
278.373 | |
BVQJQTMSTANITJ-UHFFFAOYSA-N | |
78401 | |
CCCCCCCCCCCCCCP(=O)(O)O |
C14H31O3P | |
MFCD00015834 | |
phosphonic acid, tetradecyl, 1-tetradecanephosphonic acid, tetradecanephosphonic acid, tetradecyl phosphonic acid, n-tetradecylphosphonic acid, tetradecyl-phosphonic acid, 1-tetradecylphosphonic acid, phosphonic acid, p-tetradecyl, p-tetradecyl-phosphonic acid, tetradecylphosphonate | |
tetradecylphosphonic acid |
Specifications
4671-75-4 | |
MFCD00015834 | |
Soluble in ethanol,dimethyl sulfoxide and dimethyl formamide. Partially soluble in water. | |
CCCCCCCCCCCCCCP(=O)(O)O | |
278.373 | |
278.37 | |
1-Tetradecylphosphonic acid |
C14H31O3P | |
phosphonic acid, tetradecyl, 1-tetradecanephosphonic acid, tetradecanephosphonic acid, tetradecyl phosphonic acid, n-tetradecylphosphonic acid, tetradecyl-phosphonic acid, 1-tetradecylphosphonic acid, phosphonic acid, p-tetradecyl, p-tetradecyl-phosphonic acid, tetradecylphosphonate | |
BVQJQTMSTANITJ-UHFFFAOYSA-N | |
tetradecylphosphonic acid | |
78401 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 225-118-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only