missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Trifluoromethyl)cyclopentanecarboxylic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
1-(Trifluoromethyl)cyclopentanecarboxylic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(Trifluoromethyl)cyclopentanecarboxylic acid is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
1-(Trifluoromethyl)cyclopentanecarboxylic acid is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 36°C to 38°C |
Quantity | 250 mg |
Solubility Information | Insoluble in water. |
Formula Weight | 182.15 |
Percent Purity | 97% |
Chemical Name or Material | 1-(Trifluoromethyl)cyclopentanecarboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'