Learn More
1-(Triisopropylsilyl)pyrrole, 95%, Thermo Scientific Chemicals
Description
1-(Triisopropylsilyl)pyrrole may be employed as reagent in perfluoroalkylation and Vilsmeier formylation reactions. It may be used in the preparation of ethyl 2-(2,4-dinitrophenylhydrazono]-3-[ 1-(triisopropylsily1)-pyrrol-2-yflpropanoate; heterocyclic base, 3-nitropyrrole and 3-nitropyrrole, required for the synthesis of 1 -(2?-deoxy-β-D-ribofuranosyl)-3-nitropyrrole. It participates in various electrophilic substitution reactions specifically at β-position, via reaction with various electrophilic reagents (Br+, I+,NO2+,etc).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-(Triisopropylsilyl)pyrrole may be employed as reagent in perfluoroalkylation and Vilsmeier formylation reactions. It may be used in the preparation of ethyl 2-(2,4-dinitrophenylhydrazono]-3-[ 1-(triisopropylsily1)-pyrrol-2-yflpropanoate; heterocyclic base, 3-nitropyrrole and 3-nitropyrrole, required for the synthesis of 1 -(2′-deoxy-β-D-ribofuranosyl)-3-nitropyrrole. It participates in various electrophilic substitution reactions specifically at β-position, via reaction with various electrophilic reagents (Br+, I+,NO2+,etc).
Solubility
Not miscible in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
Density | 0.904 |
Boiling Point | 78°C (0.4 mmHg) |
Flash Point | 106°C (222°F) |
Refractive Index | 1.492 |
Quantity | 5 g |
Beilstein | 3540663 |
Sensitivity | Moisture sensitive |
Solubility Information | Not miscible in water. |
Formula Weight | 223.44 |
Percent Purity | 95% |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.