missing translation for 'onlineSavingsMsg'
Learn More

1-Trityl-(R)-3-methylpiperazine, 98%, ee 99%, Thermo Scientific™

Product Code. 11364506
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313657-75-9
Molecular Formula C24H26N2
Molecular Weight (g/mol) 342.486
MDL Number MFCD03426411
InChI Key VYUQPZHRDKLOPX-HXUWFJFHSA-N
Synonym r-3-methyl-1-tritylpiperazine, r-4-n-trityl-2-methyl piperazine, 1-trityl-r-3-methylpiperazine, r-2-methyl-4-tritylpiperazine, 3r-3-methyl-1-triphenylmethyl piperazine, s-4-n-trityl-2-methylpiperazine, r-4trmp, 3r-3-methyl-1-tritylpiperazine, 3r-3-methyl-1-trityl-piperazine, r-1-trityl-3-methylpiperazine
PubChem CID 2760887
IUPAC Name (3R)-3-methyl-1-tritylpiperazine
SMILES CC1CN(CCN1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 128°C to 129°C
Quantity 250 mg
Optical Rotation −7.6° (c=1 in MeOH)
Formula Weight 342.49
Percent Purity 98%
Assay ee 99%
Chemical Name or Material 1-Trityl-(R)-3-methylpiperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'