missing translation for 'onlineSavingsMsg'
Learn More

1-Vinylimidazole, 99%

Product Code. 11302917
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11302917

Brand: Thermo Scientific Alfa Aesar L16174.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1072-63-5 | C5H6N2 | 94.12 g/mol

1-Vinylimidazole is used as a monomer in the synthesis of molecularly imprinted polymers. It is involved in the removal of vinyl protecting groups with an isoprene-catalyzed lithiation process. Also, it is involved in the preparation of magnetic-poly(divinylbenzene-1-vinylimidazole) microbeads. Further, it is used to prepare ion imprinted polyvinylimidazole-silica hybrid copolymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Vinylimidazole is used as a monomer in the synthesis of molecularly imprinted polymers. It is involved in the removal of vinyl protecting groups with an isoprene-catalyzed lithiation process. Also, it is involved in the preparation of magnetic-poly(divinylbenzene-1-vinylimidazole) microbeads. Further, it is used to prepare ion imprinted polyvinylimidazole-silica hybrid copolymer.

Solubility
Miscible with water.

Notes
Hygroscopic. Light sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-63-5
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00005297
InChI Key OSSNTDFYBPYIEC-UHFFFAOYSA-N
Synonym 1-vinylimidazole, 1-vinyl-1h-imidazole, n-vinylimidazole, 1h-imidazole, 1-ethenyl, lufixan, polyvinylimidazole, poly vinylimidazole, poly n-vinylimidazole, poly 1-vinylimidazole, n-vinylimidazole polymer
PubChem CID 66171
SMILES C=CN1C=CN=C1

Specifications

Melting Point <-50°C
Density 1.038
Boiling Point 192°C to 194°C
Flash Point 84°C (183°F)
Refractive Index 1.5315
Quantity 25 g
UN Number UN3267
Beilstein 105321
Sensitivity Hygroscopic
Solubility Information Miscible with water.
IUPAC Name 1-ethenylimidazole
Formula Weight 94.12
Percent Purity 99%
Chemical Name or Material 1-Vinylimidazole
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.