Learn More
10,11-Dihydrocarbamazepine, 99%, Thermo Scientific Chemicals
Description
10,11-Dihydrocarbamazepine is used as internal standard during simultaneous determination of carbamazepine and its active metabolite in plasma by liquid chromatography. It can be used in chemiluminescence method for determination of carbamazepine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
10,11-Dihydrocarbamazepine is used as internal standard during simultaneous determination of carbamazepine and its active metabolite in plasma by liquid chromatography. It can be used in chemiluminescence method for determination of carbamazepine.
Solubility
Soluble in dichloromethane. Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Melting Point | 204°C to 206°C |
Quantity | 250 mg |
Beilstein | 196969 |
Solubility Information | Soluble in dichloromethane. Insoluble in water. |
Formula Weight | 238.29 |
Percent Purity | 99% |
Chemical Name or Material | 10,11-Dihydrocarbamazepine |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10190416 | Certificate of Analysis | 12/09/2023 | L17271.MD,L17271.MF |
10115094 | Certificate of Analysis | 08/12/2021 | L17271.MD,L17271.MF |
10190416 | Certificate of Analysis | 06/12/2021 | L17271.MD,L17271.MF |
10030803 | Certificate of Analysis | 06/12/2021 | L17271.MD,L17271.MF |
10145392 | Certificate of Analysis | 06/12/2021 | L17271.MD,L17271.MF |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.