missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies 10% Bovine Serum Albumin in Iscove's MDM
10% BSA in Iscove's MDM
Brand: STEMCELL Technologies 09300
Additional Details : Weight : 0.10000kg
Description
- This product has been developed for use in applications where a culture medium of defined composition is required
- It contains pre-screened batches of bovine serum albumin (BSA) that have been selected to support the optimal growth of human hematopoietic progenitor cells in serum-free media formulations
- It is also suitable for the culture of mouse hematopoietic progenitor cells in serum-free conditions
- Recommended for use with MethoCult™ H4100 or MethoCult™ M3134 for hematopoietic colony-forming unit (CFU) assays (at a final concentration of 1%)
Specifications
Hematopoietic Stem and Progenitor Cells | |
Cell Culture | |
100 mL |