missing translation for 'onlineSavingsMsg'
Learn More

10-Chloro-3-decyne, 98%, Thermo Scientific™

Product Code. 11417144
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18295-64-2
Molecular Formula C10H17Cl
Molecular Weight (g/mol) 172.696
MDL Number MFCD01321154
InChI Key BQQIODGRFRSXTM-UHFFFAOYSA-N
Synonym 10-chloro-3-decyne, 3-decyne, 10-chloro, chlorodecyne, 7-decyn-1-yl chloride, acmc-209ek5, bqqiodgrfrsxtm-uhfffaoysa
PubChem CID 87561
IUPAC Name 10-chlorodec-3-yne
SMILES CCC#CCCCCCCCl

Specifications

Density 0.92
Boiling Point 122°C (20 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.462
Quantity 5 g
Formula Weight 172.7
Percent Purity 98%
Chemical Name or Material 10-Chloro-3-decyne
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'