missing translation for 'onlineSavingsMsg'
Learn More

10-Isocyanatodecyltrimethoxysilane, 95%, Thermo Scientific™

Product Code. 15406387
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310372-82-7
Molecular Formula C14H29NO4Si
Molecular Weight (g/mol) 303.474
MDL Number MFCD19105613
InChI Key GUOFKJBNQHCAKJ-UHFFFAOYSA-N
Synonym 10-isocyanatodecyl trimethoxysilane
PubChem CID 87686085
IUPAC Name 10-isocyanatodecyl(trimethoxy)silane
SMILES CO[Si](CCCCCCCCCCN=C=O)(OC)OC

Specifications

Boiling Point 120°C to 125°C (0.1mmHg)
Quantity 250 mg
Linear Formula OCN(CH2)10Si(OCH3)3
UN Number UN2810
Formula Weight 303.47
Percent Purity 95%
Sensitivity Moisture sensitive
Chemical Name or Material 10-Isocyanatodecyltrimethoxysilane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.