Learn More
Axygen™ 10 μL Microvolume Filter Tips
Axygen Universal Tips designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
Brand:  Axygen™ TF-400-R-S
 View more versions of this product
Product Code. 12555537
  • £94.00 / Pack of 960
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Technologically advanced filter material allows free passage of air while blocking aerosol contamination.
  • Filter membrane contains no leachable additives.
  • Axygen™ Maxymum Recovery™ surface improves accuracy and ensures maximum sample recovery.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Human gDNA- and PCR-inhibition-free
  • Order Info
    • Non-Filtered bulk: 1000 tips/pack, 240 packs/case, Filtered bulk: 1000 tips/pack, 10 packs/case
    • Racked: 96 tips/rack, 10 racks/pack, 5 packs/case
    • Stack Racks: 96 tips/rack, 5 racks/stack,10 stacks/case
    TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    0.5 to 10 μL
    Sterile
    USP 3127
    Allows recovery of costly samples with improved precision and accuracy
    Microvolume
    Eppendorf™ Research™ pipettes, 0.5 to 10μL
    Filtered
    Clear
    DNase, Rnase Free
    Rack
    96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
    Polypropylene