missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ML 221, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Apelin receptor (APJ) antagonist (IC50 values are 0.70 and 1.75 μM in a cAMP assay and β-arrestin assay, respectively). Displays >37-fold selectivity over the closely related angiotensin II type 1 (AT1) receptor. Exhibits no toxicity towards human hepatocytes at concentrations >50 μM.
Specifications
Quantity | 10 mg |
Formula Weight | 385.35 |
Percent Purity | >99% |
Chemical Name or Material | ML 221 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'