missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ 10% Neutral Buffered Formalin

Reduce exposure to toxic fumes with the premixed Epredia™ Neutral Buffered Formalin (10%). AFIP formulated.

Brand:  Epredia™ 5701


 View more versions of this product

Product Code. 10463750

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Phosphate buffers are used to stabilize the pH between 6.8 and 7.2
  • Stringent quality control ensures a formalin concentration of 10%
Specifications

Specifications

Formalin
1 gal.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 10% Neutral Buffered Formalin

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P235-Keep cool.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers