missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Wnt-3/sFRP-1 Complex Protein

Product Code. 18719326
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18719326

Brand: R&D Systems™ 11200WN010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Measured by its ability to activate Wnt induced TCF reporter activity in HEK293 human embryonic kidney cells. The ED50 for this effect is 7.00-63.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P17553.1, Q8C4U3.3
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS and NaCl with Trehalose.
Gene ID (Entrez) 7473
Molecular Weight (g/mol) M.W.-observed: 36-45 kDa, under reducing conditions., M.W.-theroretical: 37 kDa (Wnt-3) & 33 kDa (sFRP-1)
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse Wnt-3 protein Mouse Wnt-3 (Gly22-Lys355) (C-terminus) Accession # P17553.1 Mouse sFRP-1 (Ser32-Lys314) (C-terminus) Accession # Q8C4U3.3 (N-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias FrzA, INT4, INT4Proto-oncogene Int-4 homolog, MGC131950, MGC138321, MGC138323, proto-oncogene Wnt-3, SARP-2, sFRP1, TETAMS, wingless-type MMTV integration site family, member 3, WNT3, WNT-3 proto-oncogene protein
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Wnt3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.