Learn More
Hirschmann™ 100-150 mL Oxygen Bottle
With standard ground and chamfered glass stopper.
Brand:  Hirschmann™ 6500183
Product Code. 11702293
  • £62.50 / Pack of 2
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to Winkler
  • White graduation marks
  • Clear glass for excellent clarity
  • Precision adjusted
  • With label field
  • Individual serial number on the stopper and bottle
Specifications
Specifications
Standard ground 14/23
Yes
Yes, glass
100 to 150 mL
Glass
105 ± 10 mm