missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 100% Graphite Ferrules
Composed of a soft material that is porous to oxygen, making them suitable for most applications except GC/MS interface connections
Brand: Thermo Scientific™ 29033405
41.05 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Easy-to-use
- Form a soft grip with the column and provide a stable seal

Specifications
0.25 in. O.D. | |
For Varian, Perkin-Elmer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ 100% Graphite Ferrules > 0.25 in. O.D.; For Thermo Instruments; For Varian, Perkin-Elmer
Spot an opportunity for improvement?Share a Content Correction