Learn More
Hamilton™ 100 μL 1700 Series Gas Tight Syringe
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
Brand:  Hamilton™ CAL81056
 View more versions of this product
Product Code. 17854242
  • £344.00 / Each
Explore more special offers
Description
Description
  • This series of syringes are ideal for dispensing volumes up to 100 μL.
  • A variety of different terminations and needle options are available in this series.
Specifications
Specifications
Autoclavable
22s G
0.0059 in.
0.0283 in.
1.46 mm
0.72 mm
Gas Tight Syringe
115°C
1710
1
51 mm
Removable
PTFE
0.057 in.
0.15 mm
0.305 in.
7.75 mm
2 in.
239°F
50°F
10°C
100 μL
Sharp, Bevel, Curved, Non-coring