missing translation for 'onlineSavingsMsg'
Learn More
Learn More
100 NTU Ratio Turbidity Standard, Reagecon
Suspension of extremely accurate (± 1%) polymer microspheres with guaranteed shelf life stability even after opening the bottle.
Brand: Reagecon CRSR-100-500
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Turbidity Reference Standard, Turbidity Reference Material.
- Designed and formulated to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards, primarily formazin-based standards.
- Certified
- NIST traceable
- Non-toxic, non-carcinogenic
- Highly accurate
- Stable
- EPA approved
Specifications
Ratio Instruments | |
Accuracy ± 1% NTU | |
500 mL |
Liquid format in an HDPE Bottle GL45 | |
100 NTU ± 1% |