missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Scienceware™ 100-Place Polypropylene Storage Boxes

100-place polypropylene storage boxes provide compact, safe storage for 0.5, 1.5 and 2.0mL tubes and cryo vials. Unlike cardboard boxes they will not break down if exposed to moisture or heavy use.

Brand:  Bel-Art™ F18851-0016

 View more versions of this product

Product Code. 15446343

  • £112.00 / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Great for organization on the bench, in the freezer, or during transport
  • Can be used at temperatures -80° C to +121°C
  • Hinged lids with snap tab open easily and close securely; clear translucent color always lets you see inside
  • Each box position is labeled with an alphanumeric grid
  • ‘No slide’ features molded in bottom and lid provide secure stacking

These boxes do not fit in standard freezer racks

Order Info

Freezer racks available separately.

Specifications

Specifications

Assorted
Square
100 (0.5, 1.5 and 2 mL Tubes and Cryo Vials)
100
5/Pk.
154 mm
56 mm
Polypropylene
For 0.5, 1.5 and 2.0mL Tubes and Cryo Vials
100
Cryo Storage Box
154 x 143 x 56 mm
143 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.