Learn More
Axygen™ 1000 μL Universal Pipetter Tips: Bevelled, Maxymum Recovery
Compatible with most popular brands of pipettes
Brand:  Axygen™ T-1000-C-L-R-S
 View more versions of this product
Product Code. 12702936
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Beveled
  • Maxymum Recovery technology for ultra-low retention and high yield
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 to 1000 μL
Clear
DNase, RNase, and Pyrogen free
Low Retention, Universal
Universal Fit
Sterile
Autoclavable
Rack
Hinged rack
Polypropylene