missing translation for 'onlineSavingsMsg'
Learn More

TP-3654, MedChemExpress

Product Code. 16434536
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16434536

Brand: Medchem Express HY101126

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TP-3654 is a second-generation Pim kinase inhibitor with Ki values of 5 and 42 nM for Pim-1 and Pim-3, respectively.

TP-3654 is a second-generation Pim kinase inhibitor with Ki values of 5 and 42 nM for Pim-1 and Pim-3, respectively.

Specifications

Chemical Name or Material TP-3654
CAS 1361951-15-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H25F3N4O
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (119.49 mM; Need ultrasonic)
SMILES OC(C)(C)[C@H]1CC[C@H](NC2=NN3C(C=C2)=NC=C3C4=CC=CC(C(F)(F)F)=C4)CC1
Molecular Weight (g/mol) 418.46
Formula Weight 418.46
Percent Purity 98.48%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.