Learn More
100ML Ammonium hydroxide solution 28% NH3 in H2O,=99.99% trace metals basis
Brand: Honeywell Research Chemicals 338818-100ML
15.52 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
100ML Ammonium hydroxide solution 28% NH3 in H2O,=99.99% trace metals basis
Code : NEW
Weitere Details : CAS Number : 1336-21-6 Weight : 0.08900kg Transport : UN number : 2678 Chem class : 8 Pack group : II
Sicherheit und Handhabung
- Ammonium hydroxide solution
Signalwort
- Danger
Gefahrenkategorie
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Gefahrenhinweis
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
Sicherheitshinweis
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Ergänzende Informationen
- MIXTURE LIST-Contains : Ammonia, aqueous solution