Learn More
Endecotts™ 100 mm Diameter Stainless Steel Frame and Wire Mesh Test Sieves
Use these test sieves for all types of laboratory sampling and particle size analysis. Endecotts™ Stainless Steel Frame Test Sieves can be supplied with aperture sizes ranging from 125 mm down to 20 microns in full and half height versions.
£100.00 - £266.00
Specifications
Diameter (Metric) Outer | 100 mm |
---|---|
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
For Use With (Application) | For all types of laboratory sampling and particle size analysis |
Quantity | 1Each |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
12954205
|
Endecotts™
100SIW1.40 |
1.1 mm |
£100.00
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
12954215
|
Endecotts™
100SIW.125 |
125 μm |
£149.64
Each |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||||
12974205
|
Endecotts™
100SIW1.00 |
1 mm |
£106.00
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
12914205
|
Endecotts™
100SIW2.80 |
2.8 mm |
£100.00
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
12934215
|
Endecotts™
100SIW.250 |
250 μm |
£106.00
Each |
Estimated Shipment: 01-11-2024
Log in to see stock. |
|||||
12934205
|
Endecotts™
100SIW2.00 |
2 mm |
£106.00
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
12914225
|
Endecotts™
100SIW.032 |
32 μm |
£266.00
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
12924215
|
Endecotts™
100SIW.355 |
355 μm |
£100.00
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
12984195
|
Endecotts™
100SIW4.75 |
4.75 mm |
£113.00
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
12914215
|
Endecotts™
100SIW.425 |
425 μm |
£100.00
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
12904215
|
Endecotts™
100SIW.600 |
600 μm |
£100.00
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
12944195
|
Endecotts™
100SIW9.50 |
9.5 mm |
£113.00
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
12984215
|
Endecotts™
100SIW.053 |
53 μm |
£120.00
Each |
Estimated Shipment: 01-11-2024
Log in to see stock. |
|||||
Descripción
- Made with only the highest quality materials, available in diameter sizes of 38, 100, 150, 200, 250, 300, 315, 350, 400 or 450 mm as well as in 3, 8, 12 or 18 inches.
- Precision frame (ensures consistent nestability)
- Precise aperture (in accordance with ISO 3310-1, ASTM E11 or other specifications)
- Natural fillet (free flowing of sample)
- Totally sealed (no crevice to lose material)
- Evenly tensioned mesh (ensures accurate analysis)
- Safe edge (big radius makes it comfortable to handle)
- Serial number (ensures full traceability)
Especificaciones
100 mm | |
Stainless Steel | |
1Each | |
ISO 3310-1 & 2 requirements, ASTM, BSI |
Stainless Steel | |
For all types of laboratory sampling and particle size analysis | |
Test Sieve | |
Stainless Steel |