missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl isothiocyanate, 97%, 10g

Brand:  Thermo Scientific Maybridge TL00485EA

Cyclopentyl isothiocyanate, 97%, 10g

Code : 31

Additional Details : CAS Number : 33522-03-1 Weight : 0.06000kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III

Product Code. 10019451

  • £74.80 / 10g
Estimated Shipment: 09-10-2024
to see stock.