Learn More
Sodium HYDROXIDE 97%, 10kg
Brand: Fisher Chemical S/4880/68
171.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Sodium HYDROXIDE 97%, 10kg
Code : 21
Additional Details : CAS Number : 1310-73-2 Weight : 10.00000kg Transport : UN number : 1823 Chem class : 8 Pack group : II
Safety and Handling
- Sodium hydroxide
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.