missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HUBER 10 L Thermofluid
Recommended for many temperature control applications. Huber Kaeltmaschinenbau™ 10L Thermofluid oils provide optimal heat transfer. Temperature ranges vary from -90° – 275°C, with viscosity at 25° ranging from 3 to 50mm2/second.
£736.70 - £2522.80
Product Code | Brand | Temperature Range | Temperature (Metric) Maximum | Viscosity | Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range | Temperature (Metric) Maximum | Viscosity | Type | Price | Quantity & Availability | ||
10354295
|
HUBER
9685 |
-10°C to +120°C | 120°C | 8 mm2/sec. | Synthetic Oil |
£736.70
10 L |
Estimated Shipment: 23-10-2024
Log in to see stock. |
||
11527364
|
HUBER
6162 |
-20°C to 195°C to 235°C | 195°C open bath, 235°C closed applications (Unistats™) | 20 mm2/sec. | Silicon Oil |
£1176.60
10 L |
Estimated Shipment: 23-10-2024
Log in to see stock. |
||
11537374
|
HUBER
6158 |
20°C to 275°C | 275°C | 50 mm2/sec. | Silicon Oil |
£1123.60
10 L |
Estimated Shipment: 27-11-2024
Log in to see stock. |
||
11537364
|
HUBER
6164 |
-40°C to +165°C to 220°C | 165°C open bath, 220°C closed applications (Unistats™) | 10 mm2/sec. | Silicon Oil |
£1181.90
10 L |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||
11557364
|
HUBER
6166 |
-60°C to 115°C to 200°C | 115°C open bath, 200°C closed applications (Unistats™) | 5 mm2/sec. | Silicon Oil |
£1123.60
10 L |
Estimated Shipment: 27-11-2024
Log in to see stock. |
||
11577364
|
HUBER
6168 |
-80°C to +55°C | 55°C | 3 mm2/sec. | Silicon Oil |
£1160.70
10 L |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||
11597364
|
HUBER
6276 |
-80°C to +100°C to 250°C | 100°C open bath, 250°C closed applications (Unistats™) | 3 mm2/sec. | Silicon Oil |
£2522.80
10 L |
Estimated Shipment: 27-11-2024
Log in to see stock. |
||
11507374
|
HUBER
6259 |
-90°C to +55°C to 170°C | 55°C open bath, 170°C closed applications (Unistats™) | 3 mm2/sec. | Silicon Oil |
£2512.20
10 L |
Estimated Shipment: 27-11-2024
Log in to see stock. |
||
Description
- Thermofluids for many temperature control applications
- Display the best possible thermodynamic and environmental characteristics
- Safe reliable operation relies on compliance with safety standards to ensure optimal results
- Many varieties have higher maximum temperature when used in a Unistat™ closed application
- Convenient 10L cans of Silicon or Synthetic oil for moderate-sized storage or work spaces
Specifications
10 L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- synoil M10.120.08
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H332-Harmful if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P331-Do NOT induce vomiting.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 1-decene